missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGFBI / BIGH3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Spezifikation
| Antigen | TGFBI / BIGH3 |
|---|---|
| Konzentration | 0.23 mg/mL |
| Anwendungen | Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16846403
|
Proteintech
10188-1-AP-20UL |
20 μL |
145.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16836403
|
Proteintech
10188-1-AP-150UL |
150 μL |
410.00 €
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
This gene encodes an RGD-containing protein that binds to type I, II and IV collagens. The RGD motif is found in many extracellular matrix proteins modulating cell adhesion and serves as a ligand recognition sequence for several integrins. This protein plays a role in cell-collagen interactions and may be involved in endochondrial bone formation in cartilage. The protein is induced by transforming growth factor-beta and acts to inhibit cell adhesion. Mutations in this gene are associated with multiple types of corneal dystrophy.Spezifikation
| TGFBI / BIGH3 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P82198, Q15582 | |
| 116487, 21810, 7045 | |
| TGFBI / BIGH3 Fusion Protein Ag0241 | |
| Primary | |
| -20°C | |
| TGFBI |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Beta ig h3, BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, Kerato epithelin, LCD1, RGD CAP, TGFBI | |
| TGFBI | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts