Learn More
TH Mouse anti-Human, Clone: 1D8, Abnova™
Mouse monoclonal antibody raised against full length recombinant human TH.
Marke: Abnova MAB12883.50uL
Beschreibung
The protein encoded by this gene is involved in the conversion of tyrosine to dopamine. It is the rate-limiting enzyme in the synthesis of catecholamines, hence plays a key role in the physiology of adrenergic neurons. Mutations in this gene have been associated with autosomal recessive Segawa syndrome. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene. [provided by RefSeq]
Spezifikation
| TH | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:4000) The optimal working dilution should be determined by the end user. | |
| TYH | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 7054 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1D8 | |
| Mouse monoclonal antibody raised against full length recombinant human TH. | |
| In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide). | |
| TH | |
| Recombinant protein corresponding to full length human TH. | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.