Learn More
Heinz Herenz Thin Wooden Applicator With Cellulose Bud Standard Tip
Click to view available options
Spitzentyp:
Standard
Packungsgröße:
100 Stück
Beschreibung
We offer a wide range of sterile and nonsterile swabs:
- Wood, PS, PP, aluminium or paper applicator
- Each in various lengths available
- With cotton, viscose, polyester or foam head
- Diameter of the heads: small, medium or large
- This guarantees that the right instrument is available for every special application.
- For example, the cotton-tipped swabs are recommended for use by ENT specialists.
- Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
- Only suitable for single use
- Use by medical personnel only
Spezifikation
Spezifikation
| Autoklavierbar | Not Autoclavable |
| Sterilität | Non-sterile |
| Griffmaterial | Wood |
| Produkttyp | Applicator Stick |
| Spitzenmaterial | Wood |
| Spitzentyp | Standard |