missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thymosin alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00 € - 559.00 €
Spezifikation
| Antigen | Thymosin alpha 1 |
|---|---|
| Verdünnung | Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Anwendungen | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18462311
|
Novus Biologicals
NBP2-14801-25ul |
25ul |
369.00 €
25 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18129031
|
Novus Biologicals
NBP2-14801 |
0.1 mL |
559.00 €
0.10 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Thymosin alpha 1 Polyclonal specifically detects Thymosin alpha 1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Spezifikation
| Thymosin alpha 1 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| alpha (gene sequence 28), gene sequence 28, MGC104802, prothymosin alpha, prothymosin alpha protein, prothymosin, alpha, PTMA prothymosin, alpha, TMSA | |
| PTMA | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5757 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EVVEEAENGRDAPANGNANEENGEQEADNEVDEEEE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts