Beschreibung
Thyroid Peroxidase Monoclonal antibody specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Spezifikation
Spezifikation
| Antigen | Thyroid Peroxidase |
| Anwendungen | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Klassifikation | Monoclonal |
| Klon | TPO/1922 |
| Konjugat | PerCP |
| Zusammensetzung | PBS |
| Gen-Alias | EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant fragment of human Thyroid Peroxidase (around aa 685-804) (Exact sequence is proprietary) (Uniprot: P07202) |
| Reinigungsverfahren | Protein A or G purified |
| Mehr anzeigen |