missing translation for 'onlineSavingsMsg'
Learn More

TIA1 Antibody (rTIA1/1353), Alexa Fluor™ 594, R&D Systems™

Product Code. 18611960 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
18611960 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18611960 Supplier Novus Biologicals Supplier No. NBP321032AF594
 Eingeschränkter Artikel
Verfügbar ab: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

TIA1 Monoclonal antibody specifically detects TIA1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIA1
Anwendungen Immunohistochemistry (Paraffin)
Klassifikation Monoclonal
Klon rTIA1/1353
Konjugat Alexa Fluor 594
Zusammensetzung 50 mM Sodium Borate
Gen-Alias cytotoxic granule-associated RNA-binding protein, nucleolysin TIA-1 isoform p40, p40-TIA-1, p40-TIA-1 (containing p15-TIA-1), RNA-binding protein TIA-1, T-cell-restricted intracellular antigen-1, TIA-1, TIA1 cytotoxic granule-associated RNA binding protein, TIA1 cytotoxic granule-associated RNA-binding protein
Wirtsspezies Mouse
Immunogen Recombinant full-length human TIA1 protein
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Apoptosis, Immunology
Primär oder sekundär Primary
Gen-ID (Entrez) 7072
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Produkttyp Antibody
Form Purified
Isotype IgG1 κ
Produktlinie Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.