Learn More
Invitrogen™ TIGIT Recombinant Rabbit Monoclonal Antibody (HL2655)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA556896
Artikelnummer. 30275380
  • 514.00 € / 100 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
TIGIT Recombinant Monoclonal Antibody for Western Blot
TIGIT is a 26 kDa protein that belongs to the poliovirus receptor (PVR) family. It is expressed on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells, but not on B cells or naive CD4+ T cells. However, TIGIT can be upregulated on naive CD4+ T cells upon activation. TIGIT interacts with members of the PVR and PVR-like families, including CD155. This interaction mediates the engagement of NK and T cells with antigen-presenting cells, fibroblasts, and endothelial cells expressing PVR and PVR-like proteins. Binding of TIGIT to PVR leads to increased secretion of IL-10 and decreased secretion of IL-12B, promoting the generation of mature immunoregulatory dendritic cells and suppressing T cell activation. Through these interactions, TIGIT plays a significant role in modulating immune responses and maintaining immune homeostasis.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
TIGIT
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Q495A1
TIGIT
Recombinant protein encompassing a sequence within the Intracellular domain of human TIGIT. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL2655
Unconjugated
TIGIT
ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM
Rabbit
Protein A
RUO
201633
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen