Learn More
Invitrogen™ TIMM50 Recombinant Rabbit Monoclonal Antibody (24GB5300)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557783
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TIMM50 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TIMM50 (Mitochondrial import inner membrane translocase subunit TIM50) is an enzyme that in humans is encoded by the TIMM50 gene. Tim50 is a subunit of the Tim23 translocase complex in the inner mitochondrial membrane. Mutations in TIMM50 can lead to epilepsy, severe intellectual disability, and 3-methylglutaconic aciduria. TIMM50 expression is increased in breast cancer cells and decreased in hypertrophic hearts.
Spezifikation
| TIMM50 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TIMM50 | |
| 2810403L02Rik; AU015082; homolog of yeast Tim50; hypothetical protein LOC505489; LOC687295; LOW QUALITY PROTEIN: mitochondrial import inner membrane translocase subunit TIM50; MGC102733; mitochondrial import inner membrane translocase subunit TIM50; PRO1512; similar to translocase of inner mitochondrial membrane 50 homolog; TIM50; TIM50L; Tim50-like protein; timm50; timm50 protein; translocase of inner mitochondrial membrane 50; translocase of inner mitochondrial membrane 50 homolog; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); translocase of inner mitochondrial membrane 50-like protein; zgc:66357 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 66525, 687295, 92609 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5300 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3ZCQ8, Q9D880 | |
| TIMM50 | |
| A synthesized peptide derived from human TIM50 (aa 250-300). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |