Learn More
Invitrogen™ TIMM50 Recombinant Rabbit Monoclonal Antibody (24GB5300)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA557782
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30275761
  • 580.00 € / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
TIMM50 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TIMM50 (Mitochondrial import inner membrane translocase subunit TIM50) is an enzyme that in humans is encoded by the TIMM50 gene. Tim50 is a subunit of the Tim23 translocase complex in the inner mitochondrial membrane. Mutations in TIMM50 can lead to epilepsy, severe intellectual disability, and 3-methylglutaconic aciduria. TIMM50 expression is increased in breast cancer cells and decreased in hypertrophic hearts.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
TIMM50
Recombinant Monoclonal
Unconjugated
TIMM50
2810403L02Rik; AU015082; homolog of yeast Tim50; hypothetical protein LOC505489; LOC687295; LOW QUALITY PROTEIN: mitochondrial import inner membrane translocase subunit TIM50; MGC102733; mitochondrial import inner membrane translocase subunit TIM50; PRO1512; similar to translocase of inner mitochondrial membrane 50 homolog; TIM50; TIM50L; Tim50-like protein; timm50; timm50 protein; translocase of inner mitochondrial membrane 50; translocase of inner mitochondrial membrane 50 homolog; translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae); translocase of inner mitochondrial membrane 50-like protein; zgc:66357
Rabbit
Affinity chromatography
RUO
66525, 687295, 92609
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5300
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q3ZCQ8, Q9D880
TIMM50
A synthesized peptide derived from human TIM50 (aa 250-300).
100 μL
Primary
Human, Mouse, Rat