missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMP4 Polyclonal Antibody

Goat Polyclonal Antibody
Marke: Invitrogen™ PA5149497
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
Tissue inhibitor of metalloproteinase 4 (TIMP4) is encoded by the gene TIMP4 located on chromosome 3. It belongs to the family of TIMP genes and is known to inhibit the activity of matrix metalloproteinases. Matrix metalloproteinases (MMP) are a group of proteinases that degrade the components of extracellular matrix. TIMP4 irreversibly binds to MMPs and inactivates them with the help of zinc as a cofactor. The netrin domain of TIMP4 is involved in platelet recruitment and aggregation. Other biological functions include response to cytokine and peptide hormone stimuli and involvement with the ovulation cycle and CNS development.
Spezifikation
| TIMP4 | |
| Polyclonal | |
| Unconjugated | |
| TIMP4 | |
| Metalloproteinase inhibitor 4; TIMP 4; TIMP metallopeptidase inhibitor 4; TIMP4; TIMP-4; tissue inhibitor of metalloproteinase 4; tissue inhibitor of metalloproteinases 4 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 110595, 7079 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q99727, Q9JHB3 | |
| TIMP4 | |
| Mouse myeloma cell line NS0-derived recombinant human TIMP-4, Cys30-Pro224. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur