Beschreibung
- The upper levels are removable for washing large items.
- Equipped with a system that filters, electrically heats, and forces air with a powerful blower in the wash chamber and inside hollow instruments for excellent drying results.
- A wide range of racks, nozzles, and accessories make these machines suitable for washing and drying various laboratory glassware.
Spezifikation
Spezifikation
| Display | LCD |
| Material | Stainless Steel |
| Produkttyp | Glassware Washer with Drying System and Steam Condenser |
| Modell | Automatic Door |
| Spannung | 380/400 V |