Beschreibung
- Equipped with automatic sliding door/s and is best suited for surgical and sterilization departments.
- It can be loaded with up to 6 washing levels for a total 18 DIN baskets with a large variety of washing trolleys and accessories.
Spezifikation
Spezifikation
| Display | LCD |
| Material | Stainless Steel |
| Produkttyp | Glassware Washer with Drying System and Steam Condenser |
| Modell | Dual Automatic Sliding Doors |
| Spannung | 380/400 V |