missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TLR1 Monoclonal Antibody (285923)

Rat Monoclonal Antibody
Marca: Invitrogen™ MA560989
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Reconstitute at 0.5 mg/mL in sterile PBS.
TLR1 protein is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. The Toll-like receptor (TLR) family in mammals are comprised of a family of transmembrane proteins characterized by multiple copies of leucine rich repeats in the extracellular domain and an IL-1 receptor motif in the cytoplasmic domain. The TLRs could constitute an important and unrecognized component of innate immunity in humans. In mice, TLR1 interacts with TLR2 and co-expression of TLR1 and TLR2 enhance the NF-kappaB activation in response to a synthetic lipopeptide. They recognize the lipid configuration of the native mycobacterial lipoprotein as well as several tri-acylated lipopeptides. Different length transcripts presumably resulting from use of alternative polyadenylation site, and/or from alternative splicing, have been noted for the TLR1 gene.
Specifica
| TLR1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9EPQ1 | |
| TLR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TLR1, Ser25-Asp581. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 285923 | |
| Unconjugated | |
| TLR1 | |
| CD281; KIAA0012; rsc786; TIL; TIL. LPRS5; TLR1; toll like receptor 1; Toll/interleukin-1 receptor-like protein; toll-like receptor 1 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 21897 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto