missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
TLR3 Polyclonal specifically detects TLR3 in Human, Mouse samples. It is validated for Western Blot.
Specifica
Specifica
| Antigen | TLR3 |
| Anwendungen | Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.5 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1-3 ug/ml |
| Zusammensetzung | 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gen-Alias | CD283, CD283 antigen, toll-like receptor 3 |
| Gensymbole | TLR3 |
| Wirtsspezies | Rabbit |
| Vedi altri risultati |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?