Learn More
Novus Biologicals™ TLR7 Antibody Pack, Novus Biologicals™
Antibody Pack
Marke: Novus Biologicals™ NBP2-31097
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TLR7 antibody pack antibody specifically detects TLR7 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Spezifikation
| TLR7 | |
| Polyclonal | |
| toll-like receptor 7 | |
| 5 Vials | |
| Cytokine Research, Epigenetics, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Innate Immunity, Signal Transduction, Toll Like Receptors, Virology Bacteria and Parasites | |
| 51284 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| See individual datasheets. | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |