Learn More
Invitrogen™ TLX Recombinant Rabbit Monoclonal Antibody (24GB605)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557132
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TLX Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Nuclear receptor TLX, a NR2 hepatocyte NF4-like receptor, has been shown to affect anterior brain differentiation, retinal development, and vision. It has been suggested that TLX targets genes for RAR beta2 and Pax2 during retinal development. Homozygous TLX mutant mice are viable at birth but exhibit reduced forebrain development and are more aggressive than wild-type mice. Mice with deleted TLX locus exhibit cerebral hypoplasia, blindness, and extreme aggression (fierce (frc) phenotype). TLX expression has been documented in mouse brain and eye. ESTs have been isolated from normal human brain libraries.
Spezifikation
| TLX | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NR2E1 | |
| fc30e03; fc39f12; fierce; frc; HTll; Mtl1; Mtll; NR2D1; nr2e1; Nuclear receptor subfamily 2 group E member 1; nuclear receptor subfamily 2, group E, member 1; nuclear receptor TLX; Protein tailless homolog; tailes-related receptor; tailless; tailless homolog; TLL; Tlx; wu:fc30e03; wu:fc39f12; XTLL; zgc:100991 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21907, 684085, 7101 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB605 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q64104, Q9Y466 | |
| NR2E1 | |
| A synthesized peptide derived from human NR2E1 (aa 10-42). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |