missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM176B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Spezifikation
| Antigen | TMEM176B |
|---|---|
| Konzentration | 0.2 mg/mL |
| Anwendungen | Western Blot, Immunoprecipitation |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16859304
|
Proteintech
19825-1-AP-20UL |
20 μL |
145.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16849304
|
Proteintech
19825-1-AP-150UL |
150 μL |
410.00 €
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
TMEM176B (also known as LR8) is a transmembrane protein belongs to the MS4A family of proteins. It may play a role in the process of maturation of dendritic cells. TMEM176B was first discovered in human lung fibroblasts and is associated with human small cell lung carcinoma. Studies suggest that TMEM176B might be involved in cancer and might serve as targets for antiangiogenic therapy of cancer patients. There are two isoforms produced by alternative splicing.Spezifikation
| TMEM176B | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q3YBM2, Q9R1Q6 | |
| 28959, 65963 | |
| TMEM176B Fusion Protein Ag13918 | |
| Primary | |
| -20°C | |
| TMEM176B |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| LR8, Protein LR8, TMEM176B, transmembrane protein 176B | |
| TMEM176B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts