Learn More
TNFAIP3 Mouse anti-Human, CoraLite 594, Clone: 3A11G6, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-66695-100UL
Beschreibung
The TNFAIP3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TNFAIP3. This antibody recognizes human, rat antigen. The TNFAIP3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spezifikation
| TNFAIP3 | |
| Monoclonal | |
| 2000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Immunofluorescence | |
| 3A11G6 | |
| CoraLite 594 | |
| 100 μL | |
| 7128 | |
| IgG1 |