Learn More
Invitrogen™ TOE1 Recombinant Rabbit Monoclonal Antibody (24GB1860)
Beschreibung
TOE1 Recombinant Monoclonal Antibody for Western Blot, Flow
Inhibits cell growth rate and cell cycle. Induces CDKN1A expression as well as TGF-beta expression. Mediates the inhibitory growth effect of EGR1. Involved in the maturation of snRNAs and snRNA 3'-tail processing (PubMed:28092684). [UniProt].
Spezifikation
Spezifikation
| Antigen | TOE1 |
| Anwendungen | Flow Cytometry, Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Klon | 24GB1860 |
| Konzentration | 2 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | TOE1 |
| Gen-Zugriffsnummer | Q96GM8, Q9D2E2 |
| Gen-Alias | 4930584N22Rik; 4933424D16Rik; AI413517; hCaf1z; Target of EGR1 protein 1; target of EGR1, member 1 (nuclear); TOE1 |
| Mehr anzeigen |