Learn More
TOM20 Mouse anti-Human, Unconjugated, Clone: 1D6F5, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66777-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The TOM20 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TOM20. This antibody recognizes human antigen. The TOM20 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spezifikation
| TOM20 | |
| Monoclonal | |
| 536 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1D6F5 | |
| Unconjugated | |
| 20 μL | |
| 9804 | |
| IgG2b |