Learn More
DiversifiedBiotech TOUGH-SPOTS™ Roll Labels - 0.5 mL Tubes
Beschreibung
- Circular cryogenic labels formatted for handwriting
- Pre-cut peel off labels for tube tops with 0.5 mL
- Tough Spots™ on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers
- Tough Spots™ come in a variety of colors and sizes to ensure the right label for the right job
- Label Dimensions: 0.375 in. (9.5 mm)
Spezifikation
Spezifikation
| Klebstofftyp | Peel and Stick, Acrylic |
| Farbe | Kaffee |
| Ausführung | Semi-gloss |
| Format | Roll |
| Material | Polyolefin |
| Print Technology | Laser |
| Produkttyp | Ultra Reflective Letter Label |
| Sterilität | Non-sterile |
| Menge | 1000/Pk. |
| Länge (angloamerikanisch) | 0.375 in. |
| Mehr anzeigen |