Learn More
DiversifiedBiotech TOUGH-SPOTS™ Roll Labels - 1.5 mL Tubes (13 mm)
Click to view available options
Farbe:
Gold
Kaffee
Kastanienbraun
Lavendel
Orange
Rosa
Silber
Packungsgröße:
1000 Stück
Beschreibung
- Circular cryogenic labels formatted for handwriting
- Tough Spots™ on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers
- Tough Spots™ come in a variety of colors and sizes to ensure the right label for the right job
- Adheres to most plastics (100% virgin PP tubes recommended), glass and metals without cracking, peeling or degrading
- Flexible polyolefin material and acrylic adhesive expand and contract with temperature fluctuations
- Label Dimensions: 0.50 in. (13 mm)
- Roll of 1000
Spezifikation
Spezifikation
| Klebstofftyp | Peel and Stick, Acrylic |
| Farbe | Gold |
| Ausführung | Semi-gloss |
| Format | Roll |
| Material | Polyolefin |
| Print Technology | Laser |
| Produkttyp | Ultra Reflective Letter Label |
| Sterilität | Non-sterile |
| Länge (angloamerikanisch) | 0.50 in. |
| Breite (angloamerikanisch) | 0.50 in. |
| Mehr anzeigen |