missing translation for 'onlineSavingsMsg'
Learn More

TPPP Mouse Monoclonal Antibody, Proteintech

Artikelnummer. p-200107146 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
30235394 150 μL 150 Mikroliter
30236037 20 μL 20 Mikroliter
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Product Code. 30235394

Brand: Proteintech 680481IG150UL

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The TPPP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TPPP. This antibody recognizes human, mouse, rat, pig, rabbit, chicken antigen. The TPPP antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.

Specifications

Antigen TPPP
Anwendungen Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA
Klon 1E5F5
Konzentration 500 μg/mL
Konjugat Unconjugated
Gensymbole TPPP/p25; TPPP1; p24; p25; p25alpha
Wirtsspezies Mouse
Immunogen Recombinant Protein
Reinigungsverfahren Protein A purification
Menge 150 μL
Primär oder sekundär Primary
Gen-ID (Entrez) 11076
Zielspezies Human, Mouse, Rat, Pig, Rabbit, Chicken
Inhalt und Lagerung Store at -20°C. Stable for one year after shipment.
Molekulargewicht des Antikörpers 24-29 kDa
Produkttyp Monoclonal Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.