Learn More
Invitrogen™ TPX2 Recombinant Rabbit Monoclonal Antibody (0K5B6)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Spezifikation
| Antigen | TPX2 |
|---|---|
| Klon | 0K5B6 |
| Konzentration | 1.06 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | ELISA, Western Blot |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30284412
|
Invitrogen™
MA557999 |
20 μL |
176.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
30283573
![]() |
Invitrogen™
MA558000 |
100 μL |
436.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Immunogen sequence: MSQVKSSYSY DAPSDFINFS SLDDEGDTQN IDSWFEEKAN LENKLLGKNG TGGLFQGKTP LRKANLQQAI VTPLKPVDNT YYKEAEKENL VEQSIPSNAC SSLEVEAAIS RKTPAQPQRR SLRLSAQKDL EQKEKHHVKM KAKRCATPVI IDEILPSKKM KVSNNKKKPE EEGSAHQDTA EKNASSPEKA KGRHTVPCMP PAKQKFLKST EEQELEKSMK MQQEVVEMRK KNEEFKKLAL AGIGQPVKKS.
The protein encoded by this gene is found in the nucleolus and is similar to that of a family of genes on the Y-chromosome. This gene is intronless. Defects in this gene are a cause of sudden infant death with dysgenesis of the testes syndrome (SIDDT).Spezifikation
| TPX2 | |
| 1.06 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9ULW0 | |
| 22974 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human TPX2 (NP_036244.2). | |
| Antibody |
| 0K5B6 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Tpx2 | |
| 2610005B21Rik; C20orf1; C20orf2; Differentially expressed in cancerous and non-cancerous lung cells 2; differentially expressed in lung cells; DIL2; DIL-2; FLS353; GD: HCA519; GD:C20orf1; HCA519; HCTP4; Hepatocellular carcinoma-associated antigen 519; hepatocellular carcinoma-associated antigen 90; p100; preferentially expressed in colorectal cancer; protein fls353; REPP86; restricted expression proliferation associated protein 100; restricted expression proliferation-associated protein 100; Targeting protein for Xklp2; TPX 2; Tpx2; TPX2 microtubule nucleation factor; TPX2, microtubule nucleation factor; TPX2, microtubule-associated; TPX2, microtubule-associated protein homolog; TPX2, microtubule-associated protein homolog (Xenopus laevis); TPX2, microtubule-associated, homolog | |
| Tpx2 | |
| Primary | |
| Protein A |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.