Learn More
Thermo Scientific™ Pumpenschläuche für Transferbaugruppen
Artikelnummer. 30180439 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
Anschlusstypen:
AseptiQuik G-Konnektoren
Schlauchstil:
1/2 Zoll TPE und Pumspil-Silikon
3/8 Zoll ID Silikon und Pumpsil
3/8 Zoll TPE und Pumspil-Silikon
Packungsgröße:
10 Stück
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Anschlusstypen Schlauchstil Packungsgröße
30180439 AseptiQuik G-Konnektoren 1/2 Zoll TPE und Pumspil-Silikon 10 Stück
30180436 AseptiQuik G-Konnektoren 3/8 Zoll ID Silikon und Pumpsil 10 Stück
18126331 AseptiQuik G-Konnektoren 3/8 Zoll TPE und Pumspil-Silikon 10 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180439 Lieferant Thermo Scientific™ Lieferanten-Nr. SH31342.21
  • ONLINE EXCLUSIVE PRICE
    1365.00 €
    1316.85 € / 10 Stück
    Sparen 48.15 €
    3% Rabatt
 Eingeschränkter Artikel
Verfügbar ab: 19-02-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Thermo Scientific Standardbaugruppen für den Flüssigkeitstransfer mit dem Thermo Scientific BioTitan Retentionsgerät erhöhen die Flexibilität von Einwegprodukten. Vereinfachte Konnektivität und eine höhere Prozesseffizienz für die meisten vor- und nachgelagerten Prozessabläufe.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
TRUSTED_SUSTAINABILITY
Spezifikation
Beschreibung Transfer Assembly Pump Tubing, 1/2 inch ID TPE and Pumpsil Silicone, AseptiQuik G Connectors
Inhalt und Lagerung Raumtemperatur
Menge Each
Schlauchstil 1/2 Zoll TPE und Pumspil-Silikon
Anschlusstypen AseptiQuik G-Konnektoren
Schlauchmaterial Pharmaceutical-grade thermoplastic elastomer, platinum-cured silicone