missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TRBC1 Monoclonal antibody specifically detects TRBC1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF
Specifications
Specifications
| Antigen | TRBC1 |
| Anwendungen | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) |
| Klassifikation | Monoclonal |
| Klon | JOVI.1 |
| Konjugat | DyLight 650 |
| Zusammensetzung | 50 mM Sodium Borate |
| Gen-Alias | BV05S1J2.2, TCRBC1, TRBC1 T cell receptor beta constant 1 |
| Wirtsspezies | Mouse |
| Immunogen | Human HA1.7 TCR beta chain expressed on transgenic mouse cells. |
| Reinigungsverfahren | Protein G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?