Learn More
Skytec™ TRC715 Safety Gloves
Artikelnummer. p-200110066
Click to view available options
Größe:
Small
Mittel
Groß
Extragroß
Größe 2X Large
Packungsgröße:
10 Paare
Artikelnummer. 30274509
Marke: Skytec™ SKG00016ED
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Outstanding cut resistance allowing protection in high risk applications. Breathable, long lasting and flexible glove providing excellent grip in oily conditions.
  • Recycled Polyester
  • HPPE on the inside provides comfort and protection
  • Fully encapsulated steel with recycled polyester on the outside strengthens the protection against burrs and sharp edges
  • Micro foam nitrile coating gives exceptional grip while allowing warm air and moisture from inside to escape
Spezifikation
Zertifizierungen/Konformität (Europa) CE II, UKCA, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 4X43E, EN 388:2016 + A1:2018 4X43E
Coating Material Foam Nitrile
Farbe Green, Black
Cut Resistance Rating Level E
Hand Ambidextrous
Latex Free Yes
Material Steel Fiber
Produkttyp Cut-resistant Gloves
Größe Small
Norm EN 388 4X43E
Auskleidungsmaterial HPPE, Recycled Polyester
Länge (metrisch) 230 mm