missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CPS1 Monoclonal antibody specifically detects CPS1 in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | CPS1 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | CPS1/8416 |
| Konjugat | APC |
| Zusammensetzung | PBS |
| Gen-Alias | carbamoyl-phosphate synthase [ammonia], mitochondrial, carbamoyl-phosphate synthase 1, mitochondrial, carbamoyl-phosphate synthetase 1, mitochondrial, carbamoylphosphate synthetase I, Carbamoyl-phosphate synthetase I, CPSase I, CPSASE1, EC 6.3.4.16 |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant human CPS1 protein |
| Reinigungsverfahren | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?