missing translation for 'onlineSavingsMsg'
Learn More

Triethylmethylammoniumchlorid, TRC

Product Code. 30457248
Change view
Click to view available options
Menge:
2.5 g
Unit Size:
2.50 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
30457248 2.5 g 2.50 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457248 Supplier Toronto Research Chemicals Supplier No. TRCT7766682.5G
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Triethylmethylammonium Chloride
CAS 10052-47-8
Summenformel C7H18ClN
Menge 2.5 g
InChI-Formel InChI=1S/C7H18N.ClH/c1-5-8(4,6-2)7-3;/h5-7H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].CC[N+](C)(CC)CC
IUPAC-Name Triethyl(methyl)azanium;chlorid
Molekulargewicht (g/mol) 151.68
Formelmasse 151.1128
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.