missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM21 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 11396113
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
Artikelnummer. Menge unitSize
11396113 100 μg 100 Mikrogramm
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 11396113

Marke: Invitrogen™ PA518147

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene encodes a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The encoded protein is part of the RoSSA ribonucleoprotein, which includes a single polypeptide and one of four small RNA molecules. The RoSSA particle localizes to both the cytoplasm and the nucleus. RoSSA interacts with autoantigens in patients with Sjogren syndrome and systemic lupus erythematosus. Alternatively spliced transcript variants for this gene have been described but the full-length nature of only one has been determined.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TRIM21
Anwendungen Flow Cytometry, Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.5 mg/mL
Konjugat Unconjugated
Zusammensetzung TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gen TRIM21
Gen-Zugriffsnummer P19474
Gen-Alias 52 kDa ribonucleoprotein autoantigen Ro/SS-A; 52 kDa Ro protein; 52kD Ro/SSA autoantigen; E3 ubiquitin-protein ligase TRIM21; RING finger protein 81; RING-type E3 ubiquitin transferase TRIM21; RNF81; Ro(SS-A); Ro/SSA; RO52; Sicca syndrome antigen A; sjoegren syndrome type A antigen; Sjogren syndrome antigen A1; Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro); SSA; SS-A; Ssa1; TRIM21; tripartite motif containing 21; tripartite motif protein 21; tripartite motif-containing 21; tripartite motif-containing protein 21
Gensymbole TRIM21
Wirtsspezies Goat
Immunogen Synthetic peptide sequence (CPLNIGSQGSTDY) corresponding to the C-terminus amino acids of TRIM21 (aa 463-475).
Reinigungsverfahren Ammonium sulfate precipitation
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 6737
Zielspezies Human
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.