Learn More
TRIM21/SSA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke:  Novus Biologicals NBP1-37009
Artikelnummer. 18217004
  • 570.00 € / 100 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
TRIM21/SSA1 Polyclonal specifically detects TRIM21/SSA1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
TRIM21/SSA1
Polyclonal
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
TRIM21
The immunogen for this product maps to a region between residue 250 and 300 of human Sjogren syndrome antigen A1 using the numbering given in entry NP_003132.2 (GeneID 6737).
100 μL
Primary
Human
IgG
Immunoprecipitation, Western Blot
1.0 mg/mL
Immunoprecipitation 5-15 ug/mg lysate, Western Blot (Negative)
52 kDa ribonucleoprotein autoantigen Ro/SS-A, 52 kDa Ro/SSA, E3 ubiquitin-protein ligase TRIM21, EC 6.3.2.-, rhTRIM21, RNF81RING finger protein 81, ro(SS-A), RO52SS-A, Sicca syndrome antigen A, Sjoegren syndrome type A antigen, Sjogren syndrome antigen A1 (52kDa, ribonucleoprotein autoantigen SS-A/Ro), SSA1SSA, tripartite motif containing 21,52 kDa Ro protein, tripartite motif-containing 21, Tripartite motif-containing protein 21
Rabbit
Affinity Purified
RUO
6737
Store at 4C. Do not freeze.