Learn More
TRIM25 Mouse anti-Human, CoraLite 594, Clone: 1H6G1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-67314-100UL
Beschreibung
TRIM25 (tripartite motif-containing 25) is also named as EFP, Z147, ZNF147, RNF147 and belongs to the zinc finger protein family and tripartite motif (TRIM) family. It induces the Lys63-linked ubiquitination of RIG-I, which is crucial for the cytosolic RIG-I signalling pathway to elicit host antiviral innate immunity (PMID:17392790). It is also essential not only for DDX58 ubiquitination but also for DDX58-mediated interferon-production and antiviral activity in response to RNA virus infection (PMID: 17392790).
Spezifikation
| TRIM25 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Immunofluorescence | |
| 1H6G1 | |
| CoraLite 594 | |
| 100 μL | |
| 7706 | |
| Liquid |