missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM5 alpha/beta/gamma/delta/epsilon/iota Polyclonal Antibody

Beschreibung
Immunogen sequence: EKLLLFCQED GKVICWLCER SQEHRGHHTF LTEEVAREYQ VKLQAALEML RQKQQEAEEL EADIREEKAS WKTQIQYDKT NVLADFEQLR DILDWEESNE L; Positive Samples: HeLa, Jurkat, HepG2, Mouse stomach, Mouse thymus; Cellular Location: Cytoplasm, Nucleus, P-body.
TRIM5a is a 493 amino acid member of the large tripartite motif protein (TRIM) family. TRIM proteins are composed of three zinc-binding domains, a RING, a B-box 2 and a coiled-coil domain, and they use homomultimerization to identify different cell compartments. Some TRIM proteins, such as TRIM5a, also possess a carboxy-terminal B30.2 (SPRY) domain and localize to the cytoplasm. TRIM5a mediates innate intracellular retroviral resistance, which is dependent on its carboxy-terminal domain. The three variable regions of the B30.2 domain form loops on one side of the B30.2 core structure of TRIM5a which may form a binding surface for the virus. TRIM5a trimerization plays a major role in its affinity for the retroviral capsid, and in its ability to inhibit virus infection. The linker region between the coiled-coil and B30.2 domains of TRIM5a is required for this trimerization. TRIM5a blocks infection after the virus has entered the cell.
Spezifikation
Spezifikation
| Antigen | TRIM5 alpha/beta/gamma/delta/epsilon/iota |
| Anwendungen | ELISA, Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.53 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gen | TRIM5 |
| Gen-Zugriffsnummer | Q9C035 |
| Gen-Alias | EG667823; Gm8833; ring finger protein 88; RING-type E3 ubiquitin transferase TRIM5; RNF88; TRIM5; trim5a; TRIM5alpha; tripartite motif containing 5; tripartite motif containing 5 transcript variant iota; tripartite motif containing 5 transcript variant kappa; tripartite motif protein TRIM5; tripartite motif-containing 5; tripartite motif-containing protein 5 |
| Gensymbole | TRIM5 |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?