missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIM67 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA542274
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Peptide sequence: GGVCKGATVG VLLDLNKHTL TFFINGQQQG PTAFSHVDGV FMPALSLNRN Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 92%; Horse: 92%; Human: 92%; Mouse: 92%; Rabbit: 92%; Rat: 92%.
The TRIM67 gene encodes a member of the tripartite motif (TRIM) protein family, which is involved in various cellular processes including protein ubiquitination, signal transduction, and immune responses. TRIM67 contains RING, B-box, and coiled-coil domains, characteristic of TRIM proteins, which facilitate interactions with other proteins and contribute to its role in ubiquitination and cellular regulation. TRIM67 is notably expressed in the brain and has been associated with neurodevelopmental processes, particularly influencing neuronal differentiation and synapse formation. This gene plays a regulatory role in signaling pathways that are critical for neural development, and mutations or dysregulation in TRIM67 have been linked to neurodevelopmental disorders and cognitive deficits. Additionally, TRIM67's involvement in protein degradation and signal modulation may impact cellular architecture and brain function. Ongoing research seeks to elucidate TRIM67's precise mechanisms and potential therapeutic targets for neurological conditions.
Spezifikation
| TRIM67 | |
| Polyclonal | |
| Unconjugated | |
| TRIM67 | |
| D130049O21Rik; RGD1560667; TNL; Trim67; TRIM9-like protein; TRIM9-like protein TNL; tripartite motif containing 67; tripartite motif-containing 67; tripartite motif-containing protein 67 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 440730 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6ZTA4 | |
| TRIM67 | |
| Synthetic peptide directed towards the C-terminal of human TRIM67 (aa 711-760). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur