missing translation for 'onlineSavingsMsg'
Learn More

Trypsin 1/PRSS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marke:  Novus Biologicals NB600-1277

Artikelnummer. 18254253

  • 299.00 € / 0.20 Milliliter

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Description

Description

Trypsin 1/PRSS1 Polyclonal specifically detects Trypsin 1/PRSS1 in Bovine samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Trypsin 1/PRSS1
Polyclonal
Western Blot 1:500-1:3000, ELISA 1:5000-1:25000
P00760
PRSS1
Trypsin 1/PRSS1 1/PRSS1 [Bovine Pancreas] (Uniprot: P00760)
Delipidation and Defibrination
RUO
5644
Bovine
Antisera
Western Blot, ELISA
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A
Rabbit
25.79 kDa
0.2 mL
Primary
Trypsin
Store at -20°C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only