Learn More
Novus Biologicals™ Tryptophan Hydroxylase 1/TPH-1 Antibody (TPH1/7661), Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Tryptophan Hydroxylase 1/TPH-1 Monoclonal antibody specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spezifikation
Spezifikation
| Antigen | Tryptophan Hydroxylase 1/TPH-1 |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | TPH1/7661 |
| Konzentration | 0.2 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Zusammensetzung | 10 mM PBS with 0.05% BSA |
| Gen-Alias | EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 |
| Wirtsspezies | Mouse |
| Mehr anzeigen |