Learn More
TSH beta Antibody (TSHb/7001R), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-14196PE
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spezifikation
| TSH beta | |
| Monoclonal | |
| PE | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| Recombinant human TSH beta fragment (Uniprot: P01222) | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 7252 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| TSHb/7001R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |