missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSH Receptor Monoclonal Antibody (2C11)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA528136
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TSH Receptor Monoclonal Antibody for Western Blot, Flow, ELISA, IP
TSH Receptor gene is a membrane protein and a major controller of thyroid cell metabolism. The encoded protein is a receptor for thyrothropin and thyrostimulin, and its activity is mediated by adenylate cyclase. Defects in this gene are a cause of several types of hyperthyroidism. Three transcript variants encoding different isoforms have been found for this gene.
Spezifikation
| TSH Receptor | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P16473 | |
| TSHR | |
| Recombinant Human TSH receptor. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Western Blot | |
| 2C11 | |
| Unconjugated | |
| TSHR | |
| AI481368; CHNG1; hTSHR-I; hypothroid; hyt; LGR3; MGC141920; pet; seven transmembrane helix receptor; thyroid stimulating hormone receptor; thyroid stimulating hormone receptor, isoform 2; thyroid-stimulating hormone receptor; thyroid-stimulating hormone receptor variant; thyrotropin receptor; thyrotropin receptor-I, hTSHR-I; TSHR; TSH-R; TSHR; seven transmembrane domain G protein-coupled receptor; TSHRA | |
| Mouse | |
| Protein A | |
| RUO | |
| 7253 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur