missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSLP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Marke: Novus Biologicals NB110-55233-0.1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TSLP Polyclonal specifically detects TSLP in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
| TSLP | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| thymic stromal lymphopoietin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.25-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Q969D9 | |
| TSLP | |
| Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human TSLP. The immunogen is located within the last 50 amino acids of TSLP. Amino Acid Squence: ATQAMKKRRKRKVTTNK | |
| 0.1 mg | |
| Innate Immunity | |
| 85480 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur