missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSLP Recombinant Human Monoclonal Antibody (7G4)

Human Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA560680
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TSLP Recombinant Monoclonal Antibody for ELISA
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
Spezifikation
| TSLP | |
| Recombinant Monoclonal | |
| 1.45 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q969D9 | |
| TSLP | |
| Recombinant Human TSLP protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2 |
| ELISA | |
| 7G4 | |
| Unconjugated | |
| TSLP | |
| Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 85480 | |
| -20°C or -80°C if preferred | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur