Beschreibung
TTF-1 / NKX2-1 Monoclonal antibody specifically detects TTF-1 / NKX2-1 in Human samples. It is validated for Immunohistochemistry,Western Blot
Spezifikation
Spezifikation
| Antigen | TTF-1 / NKX2-1 |
| Anwendungen | Immunohistochemistry, Western Blot |
| Klassifikation | Monoclonal |
| Klon | CL14528 |
| Konjugat | FITC |
| Zusammensetzung | PBS |
| Gen-Alias | benign chorea, Homeobox protein NK-2 homolog A, homeobox protein Nkx-2.1, NK2 homeobox 1, NK-2 homolog A, NKX2ANKX2.1, Thyroid nuclear factor 1, Thyroid transcription factor 1BHC, TITF1TEBP, TTF1, TTF1BCH, TTF-1NK-2 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from P43699, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |