missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 AL TF630 00
Click to view available options
Größe:
Klein
Mittel
Groß
Extragroß
2X-Large
3X-Large
Packungsgröße:
2 Stück
Descripción
- Ventilated coverall available in sizes small to 3X.
- Cuff elastication and elastic thumb loop on inner sleeve opening.
- Integrated dissipative socks with boot flap.
- Gray.
- The protective system made of Tychem™ 6000 AL TF630 00 and the DuPont air-line belt, model 8000513914, helps protect the worker from contaminates.
- The air distribution plenum made of Tychem™ 6000 fabric distributes cool and breathable air helping to reduce heat stress.
- The simplified closure allows an easier donning procedure and can be taken off quickly thanks to an emergency egress strip.
- In addition to its top clear visor, this ventilated suit offers a non-PVC, highly chemical-resistant 180° panoramic view.
- Tychem™ 6000 AL garments are made from a lightweight and durable fabric (<500g per garment) consisting of a proprietary barrier film laminated to Tyvek™ substrate. They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
Especificaciones
Especificaciones
| Farbe | Grau |
| Zur Verwendung mit (Anwendung) | Protecting from Contaminents |
| Hood Type | Vollständig gekapselt |
| Material | Tychem |
| Größe | 3X-Large |
| Verschlusstyp | Horizontal Zipper with Double Flap Hook and Loop |
| Einwegprodukt | Single-use |
| Geschlecht | Unisex |
| Latexfrei | Yes |
| Anzahl Taschen | Keine |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido