Learn More
DuPont™ Tychem™ 6000 AL TF630 00
Artikelnummer. 17976871
Change view
Click to view available options
Größe:
Klein
Mittel
Groß
Extragroß
2X-Large
3X-Large
Unit Size:
2 Stück
Product Code. Größe unitSize
17976871 Groß 2 Stück
17966871 Klein 2 Stück
17956861 Mittel 2 Stück
17966861 Extragroß 2 Stück
17986871 2X-Large 2 Stück
17976861 3X-Large 2 Stück
6 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17976871
Brand: DuPont™ D15571040
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Tychem™ 6000 AL TF630 00 is a ventilated coverall with stitched and over-taped seams. It has a fully enclosed over hood with clear front and crown see through visor. Front closure with double flap hook and loop, single zipper closing to center, and a horizontal opening.
  • Ventilated coverall available in sizes small to 3X.
  • Cuff elastication and elastic thumb loop on inner sleeve opening.
  • Integrated dissipative socks with boot flap.
  • Gray.
  • The protective system made of Tychem™ 6000 AL TF630 00 and the DuPont air-line belt, model 8000513914, helps protect the worker from contaminates.
  • The air distribution plenum made of Tychem™ 6000 fabric distributes cool and breathable air helping to reduce heat stress.
  • The simplified closure allows an easier donning procedure and can be taken off quickly thanks to an emergency egress strip.
  • In addition to its top clear visor, this ventilated suit offers a non-PVC, highly chemical-resistant 180° panoramic view.
  • Tychem™ 6000 AL garments are made from a lightweight and durable fabric (<500g per garment) consisting of a proprietary barrier film laminated to Tyvek™ substrate. They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
TRUSTED_SUSTAINABILITY
Specifications
Farbe Grau
Zur Verwendung mit (Anwendung) Protecting from Contaminents
Hood Type Vollständig gekapselt
Material Tychem
Größe Groß
Verschlusstyp Horizontal Zipper with Double Flap Hook and Loop
Einwegprodukt Single-use
Geschlecht Unisex
Latexfrei Yes
Anzahl Taschen Keine
Sterilität Unsteril