missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tylosin-d3, TRC
Click to view available options
Menge:
1 mg
5 mg
10 mg
Unit Size:
1 Milligramm
10 Milligramm
5 Milligramm
Specifications
Specifications
| Chemischer Name oder Material | Tylosin A D3 (N-methyl D3) |
| Hinweise zur Reinheitsqualität | HPLC |
| Summenformel | C46 H74 D3 N O17 |
| Menge | 10 mg |
| Synonym | Tylosin A D3 (N-methyl D3), [4R-(4R*,5S*,6S*,7R*,9R*,11E,13E,15R*,16R*)]-15-[[(6-Deoxy-2,3-di-O-methyl-β-D-allopyranosyl)oxy]methyl]-6-[[3,6-dideoxy-4-O-(2,6-dideoxy-3-C-methyl-α-L-ribo-hexopyranosyl)-3-(methyl-(methyl-d3)-amino)-β-D-glucopyranosyl]oxy]-16-ethyl-4-hydroxy-5,9,13-trimethyl-2,10-dioxooxacyclohexadeca-11,13-diene-7-acetaldehyde, Tylosin-D3 (N-methyl-D3), Tylosin-d3 |
| InChI-Formel | InChI=1S/C46H77NO17/c1-13-33-30(22-58-45-42(57-12)41(56-11)37(52)26(5)60-45)18-23(2)14-15-31(49)24(3)19-29(16-17-48)39(25(4)32(50)20-34(51)62-33)64-44-38(53)36(47(9)10)40(27(6)61-44)63-35-21-46(8,55)43(54)28(7)59-35/h14-15,17-18,24-30,32-33,35-45,50,52-55H,13,16,19-22H2,1-12H3/b15-14+,23-18+/t24-,25+,26-,27-,28+,29+,30-,32-,33-,35+,36-,37-,38-,39-,40-,41-,42-,43+,44+,45-,46-/m1/s1/i9D3 |
| SMILES | O=C(C[C@@H](O)[C@H](C)[C@@H](O[C@H]1[C@H](O)[C@@H](N(C([2H])([2H])[2H])C)[C@H](O[C@@H]2O[C@@H](C)[C@H](O)[C@@](O)(C)C2)[C@@H](C)O1)[C@@H](CC=O)C[C@H]3C)O[C@H](CC)[C@@H](CO[C@@H]4O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]4OC)/C=C(C)/C=C/C3=O |
| IUPAC-Name | 2-[(4R,5S,6S,7R,9R,11E,13E,15R,16R)-6-[(2R,3R,4R,5S,6R)-5-[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyl-tetrahydropyran-2-yl]oxy-3-hydroxy-6-methyl-4-[methyl(trideuteriomethyl)amino]tetrahydropyran-2-yl]oxy-16-ethyl-4-hydroxy-15-[[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyl-tetrahydropyran-2-yl]oxymethyl]-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde |
| Molekulargewicht (g/mol) | 919.11857 |
| Formelmasse | 918.538 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction