missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tyrosinase Monoclonal Antibody (T311)

Mouse Monoclonal Antibody
Marke: Invitrogen™ 356000
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Tyrosinase Monoclonal Antibody for Western Blot, IHC
The enzyme encoded by this gene catalyzes the first 2 steps, and at least 1 subsequent step, in the conversion of tyrosine to melanin. The enzyme has both tyrosine hydroxylase and dopa oxidase catalytic activities, and requires copper for function. Mutations in this gene result in oculocutaneous albinism, and nonpathologic polymorphisms result in skin pigmentation variation. The human genome contains a pseudogene similar to the 3' half of this gene.
Spezifikation
| Tyrosinase | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P11344, P14679 | |
| TYR | |
| Recombinant tyrosinase protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry, Western Blot | |
| T311 | |
| Unconjugated | |
| TYR | |
| albino; albino locus protein; ATN; C; CMM8; LB24-AB; monophenol monooxygenase; Oca1; OCA1A; OCAIA; oculocutaneous albinism IA; SHEP3; SK29-AB; skc35; Tumor rejection antigen AB; TYR; Tyrosinase; tyrosinase (oculocutaneous albinism IA); tyrosinase precursor | |
| Mouse | |
| Protein A | |
| RUO | |
| 22173, 7299 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur