Learn More
Tyrosine Hydroxylase Antibody (OTI1D8), Novus Biologicals Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Tyrosine Hydroxylase Monoclonal antibody specifically detects Tyrosine Hydroxylase in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Tyrosine Hydroxylase |
| Anwendungen | Western Blot, Immunocytochemistry/Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | OTI1D8 |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:4000, Immunocytochemistry/ Immunofluorescence 1:100 |
| Zusammensetzung | PBS (pH 7.3) 1% BSA, 50% glycerol |
| Gen-Alias | DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase |
| Wirtsspezies | Mouse |
| Immunogen | Full length human recombinant protein of human TH(NP_000351) produced in HEK293T cell. |
| Show More |