Learn More
DuPont™ Tyvek™ IsoClean™ Chemo Gown
Click to view available options
Größe:
X-Small
Small
Mittel
Groß
Extragroß
Größe 2X
3X-Large
Packungsgröße:
30 Stück
Beschreibung
- Available in sizes XS to 3X
- Increased skirt length ensures protection against frontal exposure Openness in the back closure increases wearer comfort
- Delivers an ideal balance of protection, durability and comfort
- Made of high density polyethylene using a patented flash spinning process
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based liquid splash
- Manufactured in a controlled environment
Spezifikation
Spezifikation
| Größe | Extragroß |
| Produkttyp | Controlled Environments Gown |
| Material | Polyethylene |
| Verschlusstyp | Hook and Loop |
| Einwegprodukt | Single-use |
| Sterilität | Unsteril |
| Stulpenform | Knit |
| Latexfrei | Yes |
| Ärmelausführung | Lange Ärmel |
| Autoklavierbar | Autoclavable |