Learn More
Invitrogen™ UBB (mutant) Monoclonal Antibody (83456)
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  Invitrogen™ MA561756
Artikelnummer. 30534048
  • 606.00 € / 500 Mikrogramm
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
UBB (ubiquitin B) is a gene that encodes ubiquitin, a small regulatory protein that is ubiquitously expressed in eukaryotic cells. Ubiquitin plays a critical role in various cellular processes, including protein degradation, DNA repair, cell cycle regulation, and response to oxidative stress, by tagging proteins for degradation via the ubiquitin-proteasome system. Mutations in the UBB gene can lead to the production of defective ubiquitin proteins that can disrupt these essential cellular processes. One notable mutant form, UBB+1, is produced through a molecular process called frameshift mutation, which can occur during transcription. This mutant ubiquitin acts as a proteasome inhibitor rather than a substrate, leading to the accumulation of misfolded or damaged proteins. Such accumulation is associated with neurodegenerative diseases, including Alzheimer's disease and other forms of dementia, where impaired protein degradation contributes to disease pathology. Understanding the impact of UBB mutations, particularly UBB+1, is crucial for developing therapeutic strategies aimed at enhancing proteasome function and mitigating the effects of protein aggregation in neurodegenerative disorders. Research into UBB mutants also provides broader insights into the mechanisms of protein homeostasis and the potential for targeting the ubiquitin-proteasome system in various diseases.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
UBB (mutant)
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
E. coli-derived recombinant human Ubiquitin+1, Met1-Gly75, YADLREDPDRQDHHPGSGAQ (C-terminal sequence generated by a frameshift).
500 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
ELISA, Western Blot
83456
Unconjugated
Mouse
Protein A/G
RUO
Human
Antibody
IgG2b
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen