Learn More
UBE3B Antibody (OTI4E4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP3-44686
Artikelnummer. 30306411
  • 735.00 € / 100 Mikrogramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
UBE3B Monoclonal antibody specifically detects UBE3B in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
UBE3B
Monoclonal
Unconjugated
Lyophilized powder ( 1X PBS, pH 7.3,)
Mouse
>95%
RUO
89910
Human, Mouse, Rat
Antibody
IgG1
Western Blot
OTI4E4
Western Blot 1:2000
DKFZp586K2123, DKFZp686A1051, EC 6.3.2, EC 6.3.2.-, FLJ45294, MGC131858, MGC78388, ubiquitin protein ligase E3B, ubiquitin-protein ligase E3B
Human recombinant protein fragment corresponding to amino acids 768-1068 of human UBE3B (NP_569733) produced in E.coli.
100 μg
Primary
For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process.
Store at -20 degrees C. Avoid freeze/thaw cycles.
Lyophilized