missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UNC84A Recombinant Rabbit Monoclonal Antibody (24GB2575)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557748
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
UNC84A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.
Spezifikation
| UNC84A | |
| Recombinant Monoclonal | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94901 | |
| SUN1 | |
| A synthesized peptide derived from human SUN1 (aa 110-167). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2575 | |
| Unconjugated | |
| SUN1 | |
| 4632417G13Rik; 5730434D03Rik; KIAA0810; mKIAA0810; Protein unc-84 homolog A; Sad1 and UNC84 domain containing 1; Sad1 unc-84 domain protein 1; sad1/unc-84 protein-like 1; SUN domain-containing protein 1; Sun1; unc-84 homolog A; Unc84a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23353 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur